missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ret, Mouse anti-Human, Clone: RET/2663, Novus Biologicals™
Mouse Monoclonal Antibody
257.00 € - 489.00 €
Specifications
| Antigen | Ret |
|---|---|
| Clone | RET/2663 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18298097
|
Novus Biologicals
NBP2-79761-20UG |
20 μg |
257.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18297456
|
Novus Biologicals
NBP2-79761-100UG |
100 μg |
489.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ret Monoclonal specifically detects Ret in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Ret | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| RET | |
| IgG2b κ | |
| Protein A or G purified |
| RET/2663 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P07949, P07949 | |
| 5979 | |
| Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title