missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Aluminum Cups for Pellet Press PP 25

Product Code. 11728224 Shop All Retsch Products
Click to view available options
Diameter (Metric):
32 mm
40 mm
Includes:
1000 pieces
600 pieces
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11728224

Brand: RETSCH 22.005.0002

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Type Aluminum Cup
Diameter (Metric) 40 mm
For Use With (Equipment) Pellet Press PP 25
Includes 600 pieces
Material Aluminum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.