Learn More
RETSCH Fluid Bed Dryer TG 200
Product Code. 11722305 Shop All Retsch Products
Change view
Click to view available options
Temperature Range (Metric):
40°C to 150°C
Unit Size:
Each
Product Code. Temperature Range (Metric) Unit Size
11722305 40°C to 150°C Each
1 options
Product Code. 11722305 Supplier RETSCH Supplier No. 70.760.0001
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Type Fluid Bed Dryer
Temperature Range (Metric) 40°C to 150°C
Includes Comfort clamping lid with filter bag
Electrical Requirements 200/240 V 50/60 Hz