missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
For use with Retsch PM 100 and PM 200 Planetary Ball Mills.
- Safe, nonslip seating with built-in anti-rotation device and conical base centering
- With O-ring for gastight and dustproof seal
- Lid is easy to grip and open
Tekniske data
Tekniske data
| Product Type | Grinding Jar |
| Capacity (English) | 1.69 oz. |
| Capacity (Metric) | 50 mL |
| For Use With (Application) | Ideal for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying |
| For Use With (Equipment) | PM 100 and PM 200 planetary ball mill |
| Includes | Nonslip seating, built-in anti-rotation device, conical base centering, O-ring, dustproof seal |
| Material | Stainless Steel |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?