missing translation for 'onlineSavingsMsg'
Learn More

Labcon Reusable Silicone Sealing Mat for Round Hole Plates

Product Code. 16699852 Shop All Labcon Products
Change view
Click to view available options
Material:
Silicone
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
16699852 Silicone Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16699852 Supplier Labcon Supplier No. 39955200009
  • ONLINE EXCLUSIVE PRICE
    47.45 €
    45.91 € / Pack of 5
    Save 1.54 €
    3% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sealing mats for Labcon PCR plates and deep well plates with round wells

  • Sealing Matt
  • For Labcon PCR plates and deep well plates with round wells
  • Silicone
  • Freezable to -80°C
  • Made in USA

Specifications

Color Natural
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
Quantity 5 Mats/Pk.
For Use With (Application) PCR
Shipping Condition RT
Product Type Refillable
Product Line PurePlus
Temperature Range (Metric) Freezable to -80°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.