missing translation for 'onlineSavingsMsg'
Learn More

Revision 4 Analytes, High Level, SPEX CertiPrep™

Product Code. 15205786 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15205786 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15205786 Supplier SPEX Certiprep Supplier No. 5242R4
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,1-Dichloropropanone513-88-22000μg/mL
1-Chlorobutane109-69-32000μg/mL
2-Butanone78-93-32000μg/mL
2-Hexanone591-78-62000μg/mL
2-Nitropropane79-46-92000μg/mL
4-Methyl-2-pentanone108-10-12000μg/mL
Acetone67-64-12000μg/mL
Acrylonitrile107-13-12000μg/mL
Allyl chloride107-05-12000μg/mL
Carbon Disulfide75-15-02000μg/mL
Chloroacetonitrile107-14-22000μg/mL
Ether60-29-72000μg/mL
Ethyl methacrylate97-63-22000μg/mL
Hexachloroethane67-72-12000μg/mL
Iodomethane74-88-42000μg/mL
Methacrylonitrile126-98-72000μg/mL
Methyl acrylate96-33-32000μg/mL
Methyl methacrylate80-62-62000μg/mL
Methyl-tert-butyl ether1634-04-42000μg/mL
Nitrobenzene98-95-32000μg/mL
Pentachloroethane76-01-72000μg/mL
Propionitrile107-12-02000μg/mL
Tetrahydrofuran109-99-92000μg/mL
trans-1,4-Dichloro-2-butene110-57-62000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Method 524.2 Revision 4 Mix (High Level)
Name Note Method 524.2
Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Methods For US EPA Method 524.2
Technique AA, ICP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol-P and T
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Method 524.2 Revision 4 Mix (High Level)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: iodomethane, methanol

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.