missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rex-1/ZFP42 Antibody (5E11A6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
230.00 € - 578.00 €
Specifications
| Antigen | Rex-1/ZFP42 |
|---|---|
| Clone | 5E11A6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18115658
|
Novus Biologicals
NBP2-37357 |
0.1 mL |
578.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615985
|
Novus Biologicals
NBP2-37357-0.025ml |
0.025 mL |
230.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rex-1/ZFP42 Monoclonal specifically detects Rex-1/ZFP42 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Rex-1/ZFP42 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 132625 | |
| Purified recombinant fragment of human ZFP42 (AA: 249-310) expressed in E. Coli. | |
| Primary | |
| 34.8 kDa |
| 5E11A6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human, Mouse | |
| FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila) | |
| ZFP42 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title