missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rex1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148730
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Involved in the reprogramming of X-chromosome inactivation during the acquisition of pluripotency. Required for efficient elongation of TSIX, a non-coding RNA antisense to XIST. Binds DXPas34 enhancer within the TSIX promoter. Involved in ES cell self-renewal. [UniProt].
Spécification
| Rex1 | |
| Polyclonal | |
| Unconjugated | |
| ZFP42 | |
| 1700021P10Rik; 2610511M11Rik; AW050347; ELOABP1; EloA-BP1; elongin A binding protein 1; elongin A-binding protein 1; elongin-A-binding protein 1; hREX-1; KIAA1138; mREX-1; reduced expression protein 1; REX1; Rex-1; REX1 transcription factor; REX1, RNA exonuclease 1 homolog; REX1, RNA exonuclease 1 homolog (S. cerevisiae); REXO1; RNA exonuclease 1 homolog; Tceb3bp1; transcription elongation factor B polypeptide 3 binding protein 1; transcription elongation factor B polypeptide 3-binding protein 1; Zfp42; Zfp-42; ZFP42 zinc finger protein; Zfp971; Zinc finger protein 42; zinc finger protein 42 homolog; Zinc finger protein 754; zinc finger protein 971; ZNF754 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 132625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q96MM3 | |
| ZFP42 | |
| E. coli-derived recombinant human Rex-1/ZFP42, Ser2-Lys150. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu