missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REXO4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93290-0.1ml
This item is not returnable.
View return policy
Description
REXO4 Polyclonal antibody specifically detects REXO4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| REXO4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.1, Exonuclease XPMC2, hPMC2, PMC2, Prevents mitotic catastrophe 2 protein homolog, REX4, RNA exonuclease 4 homolog (S. cerevisiae), RNA exonuclease 4, Xenopus prevents mitotic catastrophe 2 homolog, Xenopus prevents mitotic catatrophe 2 homolog, XPMC2, XPMC2 prevents mitotic catastrophe 2 homolog, XPMC2 prevents mitotic catastrophe 2 homolog (Xenopus laevis), XPMC2HREX4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-240 of human REXO4 (NP_065118.2). WKALQEWLLKQKSQAPEKPLVISQMGSKKKPKIIQQNKKETSPQVKGEEMPAGKDQEASRGSVPSGSKMDRRAPVPRTKASGTEHNKKGTKERTNGDIVPERGDIEHKKRKAKEAAPAPPTEEDIWFDDVDPADIEAAIGPEAAKIARKQLGQSEGSVSLSLVKEQAFGGL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57109 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction