missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REXO4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13221
This item is not returnable.
View return policy
Description
REXO4 Polyclonal specifically detects REXO4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| REXO4 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| EC 3.1, Exonuclease XPMC2, hPMC2, PMC2, Prevents mitotic catastrophe 2 protein homolog, REX4, RNA exonuclease 4 homolog (S. cerevisiae), RNA exonuclease 4, Xenopus prevents mitotic catastrophe 2 homolog, Xenopus prevents mitotic catatrophe 2 homolog, XPMC2, XPMC2 prevents mitotic catastrophe 2 homolog, XPMC2 prevents mitotic catastrophe 2 homolog (Xenopus laevis), XPMC2HREX4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 57109 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| REXO4 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: QEWLLKQKSQAPEKPLVISQMGSKKKPKIIQQNKKETSPQVKGEEMPAGKDQEASRGSVPSGSKMDRRAPVPRTKASGTEHNKKGTKERTNGDIVPERGDIEHKKRKAKEAAPAPPTEEDIWFDDVDP | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction