missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RFC2 Recombinant Rabbit Monoclonal Antibody (K01_3K34)
GREENER_CHOICE

Product Code. 30826308
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826308 100 μL
30830382 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826308 Supplier Invitrogen™ Supplier No. MA564348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RFC2 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of the activator 1 small subunits family. The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins, proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). Replication factor C, also called activator 1, is a protein complex consisting of five distinct subunits. This gene encodes the 40 kD subunit, which has been shown to be responsible for binding ATP and may help promote cell survival. Disruption of this gene is associated with Williams syndrome. Alternatively spliced transcript variants encoding distinct isoforms have been described. A pseudogene of this gene has been defined on chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3K34
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene RFC2
Gene Accession No. P35250, Q641W4, Q9WUK4
Gene Alias 2610008M13Rik; 40kDa; A1 40 kDa subunit; Activator 1 40 kDa subunit; activator 1 subunit 2; Activator 1 subunit C2; AI326953; MGC3665; Recc2; replication factor C (activator 1) 2; replication factor C (activator 1) 2, 40kDa; replication factor C 40 kDa subunit; Replication factor C subunit 2; replication factor C, 40 kDa; RF-C 40 kDa subunit; RFC2; RFC40
Gene Symbols RFC2
Host Species Rabbit
Immunogen Peptide.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116468, 19718, 5982
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.