missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFX1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00 € - 468.00 €
Specifications
| Antigen | RFX1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RFX1 Polyclonal antibody specifically detects RFX1 in Human samples. It is validated for Western BlotSpecifications
| RFX1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS (pH 7.3), 50% glycerol | |
| 5989 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EFC, EF-C, Enhancer factor C, MHC class II regulatory factor RFX, MHC class II regulatory factor RFX1, Regulatory factor X 1, regulatory factor X, 1 (influences HLA class II expression), RFX, trans-acting regulatory factor 1, Transcription factor RFX1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human RFX1 (NP_002909.4). QQVHSPPEQSPVQANSSSSKTAGAPTGTVPQQLQVHGVQQSVPVTQERSVVQATPQAPKPGPVQPLTVQGLQPVHVAQEVQQLQQVPVPHVYSSQVQYVEG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title