missing translation for 'onlineSavingsMsg'
Learn More

RG108, MedChemExpress

Product Code. 16421947
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16421947

Brand: Medchem Express HY13642

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RG108 (N-Phthalyl-L-tryptophan) is a non-nucleoside DNA methyltransferases (DNMTs) inhibitor (IC50=115 nM) that blocks the DNMTs active site. RG108 (N-Phthalyl-L-tryptophan) causes demethylation and reactivation of tumor suppressor genes, but it does not affect the methylation of centromeric satellite sequences.

RG108 (N-Phthalyl-L-tryptophan) is a non-nucleoside DNA methyltransferases (DNMTs) inhibitor (IC50=115 nM) that blocks the DNMTs active site. RG108 (N-Phthalyl-L-tryptophan) causes demethylation and reactivation of tumor suppressor genes, but it does not affect the methylation of centromeric satellite sequences.

Specifications

Chemical Name or Material RG108
Physical Form Powder
CAS 48208-26-0
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H14N2O4
For Use With (Application) Cancer-programmed cell death
Synonym N-Phthalyl-L-tryptophan
Solubility Information DMSO : 100 mg/mL (299.11 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N1[C@H](C(O)=O)CC2=CNC3=C2C=CC=C3)C4=CC=CC=C4C1=O
Molecular Weight (g/mol) 334.33
Formula Weight 334.33
Percent Purity 99.51%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.