missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGFP966, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
RGFP966 is a highly selective HDAC3 inhibitor with an IC50 of 80 nM and shows no inhibition to other HDACs at concentrations up to 15 μM. RGFP966 can penetrate the blood brain barrier (BBB).
Specifications
Specifications
| Chemical Name or Material | RGFP966 |
| CAS | 1357389-11-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H19FN4O |
| Quantity | 100 mg |
| Solubility Information | DMF : 50 mg/mL (137.97 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (137.97 mM; Need ultrasonic) |
| SMILES | O=C(NC1=CC=C(F)C=C1N)/C=C/C2=CN(C/C=C/C3=CC=CC=C3)N=C2.[E] |
| Molecular Weight (g/mol) | 362.4 |
| Formula Weight | 362.4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction