Learn More
RGM-A Antibody (CL11342) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08000G
Product Code. 30292601
  • 737.00 € / 0.10mL
Description
RGM-A Monoclonal antibody specifically detects RGM-A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RGM-A
Monoclonal
DyLight 488
RGM domain family member A, RGM domain family, member A, RGMrepulsive guidance molecule A
This antibody was developed using a synthetic peptide derived from Q96B86, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience, Stem Cells
56963
Store at 4⪚C in the dark.
IgG2b
Western Blot
CL11342
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified