missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS2 Recombinant Rabbit Monoclonal Antibody (HL2987)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556861
This item is not returnable.
View return policy
Description
RGS2 Recombinant Monoclonal Antibody for IHC (P)
Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 2 belongs to this family. The protein acts as a mediator of myeloid differentiation and may play a role in leukemogenesis.
Specifications
| RGS2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O08849, Q9JHX0 | |
| RGS2 | |
| Recombinant protein of human RGS2. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| HL2987 | |
| Unconjugated | |
| RGS2 | |
| cell growth-inhibiting gene 31 protein; cell growth-inhibiting protein 31; G0 to G1 switch regulatory 8, 24kD; G0/G1 switch regulatory protein 8; G0S8; GIG31; GOS8; regulator of G protein signaling 2; regulator of G protein signaling 2; regulator of G-protein signaling 2; regulator of G-protein signaling 2; regulator of G-protein signaling 2, 24kDa; regulator of G-protein signaling protein 2; regulator of G-protein signalling 2, 24kDa; Rgs2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 19735, 84583 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction