missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGX-104 hydrochloride, MedChemExpress
Description
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Specifications
Specifications
| Chemical Name or Material | RGX-104 hydrochloride |
| CAS | 610318-03-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C34H34Cl2F3NO3 |
| Quantity | 2 mg |
| Solubility Information | DMSO : 150 mg/mL (237.14 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1.Cl |
| Molecular Weight (g/mol) | 632.54 |
| Formula Weight | 632.54 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?