missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHBG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526978
This item is not returnable.
View return policy
Description
RHBG Polyclonal Antibody for Western Blot, IHC (P), Flow
RHBG encodes one of two non-erythroid members of the Rhesus (Rh) protein family. Non-erythroid Rh protein family members are mainly expressed in the kidney and belong to the methylammonium-ammonium permease/ammonia transporters superfamily. All Rh family proteins are predicted to be transmembrane proteins with 12 membrane spanning domains and intracytoplasmic N- and C-termini.
Specifications
| RHBG | |
| Polyclonal | |
| Unconjugated | |
| RHBG | |
| Ammonium transporter Rh type B; Rh family B glycoprotein; Rh family B glycoprotein (gene/pseudogene); rh family type B glycoprotein; Rh family, B glycoprotein; Rh type B glycoprotein; Rhbg; Rhesus blood group family type B glycoprotein; Rhesus blood group, B glycoprotein; Rhesus blood group-associated B glycoprotein; SLC42A2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 57127, 58176 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8BUX5, Q9H310 | |
| RHBG | |
| KLH conjugated synthetic peptide between 423-450 amino acids from the C-terminal region of human RHBG. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction