missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHD Antibody (D7C2) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-09042-0.025mg
This item is not returnable.
View return policy
Description
RHD Monoclonal antibody specifically detects RHD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| RHD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| D7C2 | |
| Unconjugated | |
| blood group Rh(D) polypeptide, CD240D, DIIIc, RH, Rh blood group, D antigen, RH30, Rh4, RHCED, RhDCw, RHDel, RHDVA(TT), RhII, RhK562-II, RhPI, RHPII, RHXIII | |
| A Rh- volunteer blood donor (r/r) was injected with 5ml of Rh+ erythrocytes (R1/r) and the peripheral lymphocytes were isolated and transformed with 1 mL of culture supernatant of the EBV marmoset B96-8 cell line. | |
| 0.025 mg | |
| Cardiovascular Biology, CD Markers, Immunology | |
| 6007 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction