Learn More
RHD Antibody (LFB Anti-RhD), PerCP, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-27995PCP
Product Code. 30114143
  • 419.00 € / 0.10mL
Description
RHD Monoclonal antibody specifically detects RHD in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RHD
Monoclonal
PerCP
blood group Rh(D) polypeptide, CD240D, DIIIc, RH, Rh blood group, D antigen, RH30, Rh4, RHCED, RhDCw, RHDel, RHDVA(TT), RhII, RhK562-II, RhPI, RHPII, RHXIII
RHD/ CD240d
0.1 mL
Cardiovascular Biology, CD Markers, Immunology
6007
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
LFB Anti-RhD
PBS
Human
Protein A purified
RUO
Primary
Human
Purified