missing translation for 'onlineSavingsMsg'
Learn More

Rhenium pentacarbonyl chloride, 98%

Product Code. 10202942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10202942 5 g Glass bottle 5g
10389410 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10202942

Brand: Thermo Scientific Acros 316310050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14099-01-5 | C5ClO5Re | 361.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14099-01-5
Molecular Formula C5ClO5Re
Molecular Weight (g/mol) 361.71
MDL Number MFCD00013296
InChI Key JQUUAHKBIXPQAP-UHFFFAOYSA-M
Synonym pentacarbonylchlororhenium, carbon monoxide; chlororhenium, pentakis carbon monoxide ; chlororhenium, 5co.clre, rhenium i pentacarbonyl chloride, pentacarbonylchlororhenium i
PubChem CID 6096982
IUPAC Name carbon monoxide;chlororhenium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].Cl[Re]

Specifications

Color White
Elemental Analysis 97.5% min.
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 361.71
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material Rhenium pentacarbonyl chloride, 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.