missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
222.00 € - 561.00 €
Specifications
| Antigen | RhoA |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18266834
|
Novus Biologicals
NB100-91273-0.025ML |
0.025 mL |
222.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18740524
|
Novus Biologicals
NB100-91273 |
0.1 mL |
561.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RhoA Polyclonal specifically detects RhoA in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RhoA | |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, mTOR Pathway | |
| P61586 | |
| 387 | |
| IgG | |
| Unpurified | |
| RhOA. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA | |
| RHOA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 22 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title