missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine B, pure

Product Code. 10396870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396870 100 g Glass bottle 100g
10573871 500 g Glass bottle 500g
13408358 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10396870 Supplier Thermo Scientific Chemicals Supplier No. 132311000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
Synonym Basic Violet 10, C.I. 45170
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Absorbance 1700 (at lambda min.)
Melting Point 210.0°C to 211.0°C
Absorption Ratio 1.1 to 1.6
Beilstein 19, V,8, 669
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in hcl and naoh, soluble in alcohol
Infrared Spectrum Authentic
Lambda Max 549 to 558nm (in 50:50 IMS:water)
Formula Weight 479.01
Loss on Drying 10% max. (105°C)
Grade Pure
Packaging Glass bottle
Color Green
Quantity 100 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Rhodamine B
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Rhodamine B
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.