accessibility menu, dialog, popup

UserName

ClCH3NCH3OOHONH3CH3C(?)

CAS RN 81-88-9

IUPAC Name: 9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
Synonyms: rhodamine
Molecular Weight (g/mol): 479.02
Molecular Formula: C28H31ClN2O3
InChi Key: PYWVYCXTNDRMGF-UHFFFAOYSA-N
SMILES: [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC
Molecular Weight (g/mol) 
  • (3)
  • (12)
Quantity 
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Color 
  • (9)
Grade 
  • (8)
Percent Purity 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (11)
  • (3)
  • (3)
  • (2)
  • (3)
  • (12)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (9)
  • (8)
  • (4)
  • (1)
  • (2)