missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) nitrate solution, Rh 10-15% w/w (cont. Rh)

Product Code. 11490300
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11490300

Brand: Thermo Scientific Alfa Aesar 012633.NA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 10139-58-9 | N3O9Rh | 288.92 g/mol

It acts as a chemical catalyst, it is used as a slurry bed in Hydrogenation reaction. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10139-58-9
Molecular Formula N3O9Rh
Molecular Weight (g/mol) 288.92
MDL Number MFCD00011202
InChI Key VXNYVYJABGOSBX-UHFFFAOYSA-N
Synonym rhodium nitrate, rhodium iii nitrate, rhodium trinitrate, nitric acid, rhodium 3+ salt, rhodium 3+ trinitrate, nitric acid, rhodium 3+ salt 3:1, nitric acid,rhodium 3+ salt 3:1, rhodiumtrinitrate, rhodium +3 nitrate, acmc-20ajsk
PubChem CID 150190
IUPAC Name rhodium(3+);trinitrate
SMILES [Rh+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Red to Brown
Linear Formula Rh(NO3)3
UN Number UN3093
Concentration Rh 10 to 15% w/w (cont. Rh)
Solubility Information Soluble in alcohol,water,acetone
Formula Weight 288.92
Odor Acidic
Quantity 2 g
Physical Form Liquid
Chemical Name or Material Rhodium(III) nitrate solution

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.