missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodopsin Antibody (Rho 1D4), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81059H
This item is not returnable.
View return policy
Description
Rhodopsin Monoclonal specifically detects Rhodopsin in Human, Mouse, Rat, Amphibian, Bovine, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Rhodopsin | |
| Monoclonal | |
| HRP | |
| PBS | |
| RHO | |
| Bleached bovine rod outer segment (ROS). | |
| 0.1 mL | |
| GPCR, Phospho Specific, Signal Transduction, Vision | |
| 6010 | |
| Human, Mouse, Rat, Amphibian, Bovine, Zebrafish | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| Rho 1D4 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody binds to the C-terminus of rhodopsin on the cytoplasmic side of disk membranes. The epitope has the following amino acid sequence: 'TETSQVAPA'. A limitation of the 1D4 tag, however, is that it has to be placed at the C-terminus of a protein. This is because the Rho1D4 monoclonal antibody requires a free carboxylate group for high affinity binding. Amidation of the carboxyl group lowers its immunoreactivity to the Rho1D4 antibody by over 100 fold. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction