Learn More
RhoH Antibody (OTI9F4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44854
Product Code. 30306233
  • 735.00 € / 100µg
Description
RhoH Monoclonal antibody specifically detects RhoH in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RhoH
Monoclonal
Unconjugated
Lyophilized powder ( 1X PBS, pH 7.3,)
Mouse
>95%
RUO
Primary
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Store at -20 degrees C. Avoid freeze/thaw cycles.
Lyophilized
Western Blot
OTI9F4
Western Blot 1:2000
ras homolog gene family, member H, translocation three four
Human recombinant protein fragment corresponding to amino acids 97-190 of human RHOH(NP_004301) produced in E.coli.
100 μg
Signal Transduction
399
Human, Mouse, Rat
Antibody
IgG1