missing translation for 'onlineSavingsMsg'
Learn More

Rhosin hydrochloride, MedChemExpress

Product Code. 16495268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495268 10 mg 10mg
16457987 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495268 Supplier Medchem Express Supplier No. HY12646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rhosin hydrochloride is a potent, specific RhoA subfamily Rho GTPases inhibitor. Rhosin hydrochloride specifically binds to RhoA to inhibit RhoA-GEF interaction with a Kd of ∼ 0.4 uM, and does not interact with Cdc42 or Rac1, nor the GEF, LARG. Rhosin hydrochloride induces cell apoptosis. Rhosin hydrochloride promotes stress resiliency through enhancing D1-MSN plasticity and reducing hyperexcitability.

Rhosin hydrochloride is a potent, specific RhoA subfamily Rho GTPases inhibitor. Rhosin hydrochloride specifically binds to RhoA to inhibit RhoA-GEF interaction with a Kd of ∼ 0.4 uM, and does not interact with Cdc42 or Rac1, nor the GEF, LARG. Rhosin hydrochloride induces cell apoptosis. Rhosin hydrochloride promotes stress resiliency through enhancing D1-MSN plasticity and reducing hyperexcitability.

Specifications

Chemical Name or Material Rhosin hydrochloride
Physical Form Solid
CAS 1281870-42-5
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H19ClN6O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 100 mg/mL (253.25 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (42.22 mM; Need ultrasonic)
SMILES O=C(N/N=C/C1=CC=C2N=CC=NC2=C1)[C@H](N)CC3=CNC4=C3C=CC=C4.[H]Cl
Molecular Weight (g/mol) 394.86
Formula Weight 394.86
Percent Purity 99.67%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.