missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHOT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542646
This item is not returnable.
View return policy
Description
Peptide sequence: MKKDVRILLV GEPRVGKTSL IMSLVSEEFP EEVPPRAEEI TIPADVTPER Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Mitochondrial GTPase involved in mitochondrial trafficking. RHOT1 is probably involved in control of anterograde transport of mitochondria and their subcellular distribution.
Specifications
| RHOT1 | |
| Polyclonal | |
| Unconjugated | |
| RHOT1 | |
| 2210403N23Rik; AA415293; AF244542; AI834919; Arht1; C430039G08Rik; LOW QUALITY PROTEIN: mitochondrial Rho GTPase 1; MGC128572 protein; Miro1; MIRO-1; mitochondrial Rho (MIRO) GTPase 1; mitochondrial Rho GTPase 1; rac-GTP binding protein-like protein; Rac-GTP-binding protein-like protein; ras homolog family member T1; Ras homolog gene family member T1; ras homolog gene family, member T1; Rhot1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55288 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Surface Plasmon Resonance | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IXI2 | |
| RHOT1 | |
| Synthetic peptide directed towards the N-terminal of human RHOT1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction