missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOT2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93326-0.1ml
This item is not returnable.
View return policy
Description
RHOT2 Polyclonal antibody specifically detects RHOT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RHOT2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| ARHT2, C16orf39, chromosome 16 open reading frame 39, EC 3.6.5, EC 3.6.5.-, hMiro-2, MIRO-2mitochondrial Rho 2, mitochondrial Rho GTPase 2, Ras homolog gene family member T2, ras homolog gene family, member T2, RASL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 340-550 of human RHOT2 (NP_620124.1). PWGPELPRTVRTEAGRLPLHGYLCQWTLVTYLDVRSCLGHLGYLGYPTLCEQDQAHAITVTREKRLDQEKGQTQRSVLLCKVVGARGVGKSAFLQAFLGRGLGHQDTREQPPGYAIDTVQVNGQEKYLILCEVGTDGLLATSLDATCDVACLMFDGSDPKSFAHCASVYKHHYMDGQTPCLFVSSKADLPEGVAVSGPSPAEFCRKHRLPA | |
| 0.1 mL | |
| Signal Transduction | |
| 89941 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction