missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: KTLYDNYQRA VAKAGLASRW TNLGTVNAAA PAQPSTGPKT GTTQPNGQIP QATHSVSAVL QEAQRHAETS KDKKPAL Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 65%.
RIAM appears to function in the signal transduction from Ras activation to actin cytoskeletal remodeling. Suppresses insulin-induced promoter activities through AP1 and SRE. Mediates Rap1-induced adhesion.
Specifications
Specifications
| Antigen | RIAM |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | APBB1IP |
| Gene Accession No. | Q7Z5R6 |
| Gene Alias | 9930118P07Rik; amyloid beta (A4) precursor protein-binding, family B, member 1 interacting protein; amyloid beta A4 precursor protein-binding family B member 1-interacting protein; amyloid beta precursor protein binding family B member 1 interacting protein; APBB1-interacting protein 1; Apbb1ip; INAG1; Prel1; PREL-1; proline rich EVH1 ligand 1; proline-rich EVH1 ligand 1; Proline-rich protein 48; proline-rich protein 73; Prp48; Rap1-GTP-interacting adapter molecule; Rap1-GTP-interacting adaptor molecule; Rap1-interacting adaptor molecule; RARP1; RARP-1; retinoic acid-responsive proline-rich protein 1; RIAM; RIAMRARP-1; zgc:63968 |
| Gene Symbols | APBB1IP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
