missing translation for 'onlineSavingsMsg'
Learn More

Ribavirin Carboxylic Acid, TRC

Product Code. 30010036
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010036 25 mg 25mg
30007909 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010036 Supplier Toronto Research Chemicals Supplier No. TRCR41448525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39925-19-4
Molecular Formula C8 H11 N3 O6
Molecular Weight (g/mol) 245.19
Synonym 1-beta-D-Ribofuranosyl-1H-1,2,4-triazole-3-carboxylic Acid, Ribavirin Imp. A (EP)
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxylic acid
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc(n2)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H11N3O6/c12-1-3-4(13)5(14)7(17-3)11-2-9-6(10-11)8(15)16/h2-5,7,12-14H,1H2,(H,15,16)/t3-,4-,5-,7-/m1/s1
Formula Weight 245.0648
Chemical Name or Material 1-Beta-D-Ribofuranosyl-1H-1,2,4-triazole-3-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.