missing translation for 'onlineSavingsMsg'
Learn More

Ribonucleic acid from Baker's yeast

Product Code. 15493829
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493829 100 mg 100mg
15403839 500 mg 500mg
15483829 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493829 Supplier Thermo Scientific Alfa Aesar Supplier No. J61215.MC
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 63231-63-0

Ribonucleic acid (RNA) from baker?s yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ribonucleic acid (RNA) from baker′s yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.

Solubility
It is soluble in water.

Notes
Prepared by a modification of Crestfield, A.M., et al., J. Biol. Chem., 216, 185 (1955). Suitable for use as a ribonuclease substrate. Not suitable for use as a carrier in nucleic acid purification and precipitation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-63-0
MDL Number MFCD00132196
IUPAC Name Ribonucleic acid

Specifications

Color White to Brown
Quantity 100 mg
Solubility Information It is soluble in water.
Physical Form Powder
Chemical Name or Material Ribonucleic acid from Baker's yeast

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.