missing translation for 'onlineSavingsMsg'
Learn More

Rifampicin, MedChemExpress

Product Code. 16432119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16432119 1 g 1g
16410858 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16432119 Supplier Medchem Express Supplier No. HYB0272
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rifampicin is a potent and broad spectrum antibiotic against bacterial pathogens. Rifampicin has anti-influenza virus activities. Rifampicin shows anti-orthopoxvirus activity.

Rifampicin is a potent and broad spectrum antibiotic against bacterial pathogens. Rifampicin has anti-influenza virus activities. Rifampicin shows anti-orthopoxvirus activity.

Specifications

Chemical Name or Material Rifampicin
Physical Form Solid
CAS 13292-46-1
Color Orange Red
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C43H58N4O12
For Use With (Application) COVID-19-anti-virus
Synonym Rifampin Rifamycin AMP
Solubility Information DMSO : 25 mg/mL (30.38 mM; ultrasonic and warming and heat to 80°C) ∣H2O : 1 mg/mL (1.22 mM; ultrasonic and heat to 60°C)
SMILES OC(C(/C=N/N1CCN(C)CC1)=C(NC(/C(C)=C\C=C\[C@@H]([C@@H]([C@@H](C)[C@H]2O)O)C)=O)C(O)=C3C(O)=C4C)=C3C5=C4O[C@](C)(O/C=C/[C@@H]([C@H]([C@@]([C@@H]2C)([H])OC(C)=O)C)OC)C5=O
Molecular Weight (g/mol) 822.94
Formula Weight 822.94
Percent Purity 97.61%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.