missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Rifampicin ReadyMade™ Solution

Product Code. 30044020
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044020 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044020 Supplier TOKU-E™ Supplier No. R041100ML
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rifampicin ReadyMade™ Solution is provided as a sterile-filtered solution of Rifampicin in water.

  • CAS Number: 13292-46-1
  • Molecular Formula: C43H58N4O12
  • Molecular Weight: 822.94 g/mol
  • Storage Conditions: -20°C, Protect from light
  • Form: Liquid (in water with starch as a solubilizer)
  • Source: Semi-synthetic - Amycolatopsis rifamycinica
  • Spectrum: Rifampicin is a broad-spectrum antibiotic with a wide range of activity including: Gram-positive aerobic bacteria, particularly Staphylococcus spp and Rhodococcus equ. Brucella and some other fastidious organisms are susceptible but Gram-negative bacteria more generally are resistant; Gram-positive and Gram-negative anaerobic bacteria at low concentrations, including Bacteroides fragilis, Chlamydophila and Rickettsia; Mycobacterium tuberculosis (note: most other mycobacteria are resistant); some protozoa; some fungi; and poxviruses
  • Mechanism of Action: Rifampicin targets prokaryotic DNA-dependent RNA polymerases which prevent subsequent RNA transcription and protein translation. Bacterial resistance to Rifampicin is caused by mutations leading to a change in the structure of the β-subunit of RNA polymerase. Rifampicin inhibits the assembly of DNA and protein into mature virus particles. It inhibits initiation of RNA synthesis by binding to the β-subunit of RNA polymerase, which results in cell death. Rifampicin has been shown to inhibit α-synuclein fibrillation and disaggregate fibrils in a concentration-dependent manner. Rifampicin can activate pregnane X receptor (PXR), which affects cytochrome P450, and the activity of glucuronosyltransferases and P-glycoprotein. Rifampicin has been shown to enhance CYP2C-mediated metabolism, affecting compounds that are transported by P-glycoprotein and metabolized by CYP3A4

Specifications

CAS 13292-46-1
Chemical Name or Material Rifampicin
Color Red
Content And Storage -20°C, Protect from light
Description Rifampicin ReadyMade™ Solution 10 g/mL in Water
Concentration 8 to 12 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in Water
For Use With (Application) Microbiology, Plant Biology and Eukaryotic Cell Culture Applications
Product Line ReadyMade™ Solutions
Purity or Quality Grade >90%
Quantity 100 mL
Shipping Condition Ice Pack
Source Semi-synthetic
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.