missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIG-I Antibody (Alme-1), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-80055JF549
This item is not returnable.
View return policy
Description
RIG-I Monoclonal antibody specifically detects RIG-I in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RIG-I | |
| Monoclonal | |
| Janelia Fluor 549 | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 58, DEAD box protein 58, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, DKFZp686N19181, EC 3.6.4.13, FLJ13599, probable ATP-dependent RNA helicase DDX58, retinoic acid inducible gene I, Retinoic acid-inducible gene 1 protein, Retinoic acid-inducible gene I protein, RIG-1, RIG-IDKFZp434J1111, RNA helicase RIG-I | |
| Recombinant human RIG-I (aa 201-713). | |
| 0.1 mL | |
| Phospho Specific | |
| 23586 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alme-1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction