missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIG-I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
379.00 € - 593.00 €
Specifications
| Antigen | RIG-I |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217594
|
Novus Biologicals
NBP2-55599 |
100 μL |
593.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645597
|
Novus Biologicals
NBP2-55599-25ul |
25 μL |
379.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RIG-I Polyclonal specifically detects RIG-I in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Spezifikation
| RIG-I | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 58, DEAD box protein 58, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, DKFZp686N19181, EC 3.6.4.13, FLJ13599, probable ATP-dependent RNA helicase DDX58, retinoic acid inducible gene I, Retinoic acid-inducible gene 1 protein, Retinoic acid-inducible gene I protein, RIG-1, RIG-IDKFZp434J1111, RNA helicase RIG-I | |
| RIGI | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 23586 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NREFGTQKYEQWIVTVQKACMVFQMPDKDEESRICKALFLYTSHLRKYNDALIISEHARMKDALDYLKDFFSNVRAAGFDEIEQDLTQRFEEKLQELESVSRDPSNENPKLEDLCFILQEEYHLNPETITILFVK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts