Learn More
Invitrogen™ ALKBH4 Recombinant Rabbit Monoclonal Antibody (24GB6805)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556963
 View more versions of this product
Product Code. 30275083
  • 500.00 € / 100µL
Description
ALKBH4 Recombinant Monoclonal Antibody for Western Blot
Dioxygenase that mediates demethylation of actin monomethylated at 'Lys-84' (K84me1), thereby acting as a regulator of actomyosin-processes (PubMed:23673617). Demethylation of actin K84me1 is required for maintaining actomyosin dynamics supporting normal cleavage furrow ingression during cytokinesis and cell migration (PubMed:23673617). In addition to proteins, also demethylates DNA: specifically demethylates DNA methylated on the 6th position of adenine (N(6)-methyladenosine) DNA, thereby regulating Polycomb silencing. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ALKBH4
Recombinant Monoclonal
Unconjugated
ALKBH4
2010004B12Rik; ABH4; Abj4; alkB homolog 4, lysine demethylase; alkB homolog 4, lysine demthylase; alkB, alkylation repair homolog 4; alkB, alkylation repair homolog 4 (E. coli); Alkbh4; Alkylated DNA repair protein alkB homolog 4; alpha-ketoglutarate-dependent dioxygenase alkB homolog 4; DNA N6-methyl adenine demethylase ALKBH4; Lysine-specific demethylase ALKBH4; probable alpha-ketoglutarate-dependent dioxygenase ABH4; RGD1308608
Rabbit
Affinity chromatography
RUO
54784
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6805
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NXW9
ALKBH4
Recombinant protein of human ALKBH4 (aa 2-302).