missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ 384-Well Rigid Frame PCR Plates, BIO-CERT™ PCR QUALITY

Product Code. 17821262 Shop All Brand Products
Change view
Click to view available options
Color:
Frosted
Well Color:
Transparent
White
Unit Size:
Pack of 50
Product Code. Color Well Color unitSize
17821262 Frosted White Pack of 50
17881252 Frosted Transparent Pack of 50
2 options
This item is not returnable. View return policy

Product Code. 17821262

Brand: BRANDTECH™ BRAND™ 781539

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

384-well PCR plates are an economical solution for automatic, high-throughput analyses.

BRAND PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:

    Exceptional Quality and Purity
  • BIO-CERT™ PCR QUALITY guarantees plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
  • Manufactured without contaminants that could interfere with reagents or samples
  • Ultra-smooth vessel interiors minimize enzyme and nucleic acid binding for reliable results
    Robust Design and Durability
  • Strong polycarbonate frames provide safe handling in both semi-automated and automated pipetting systems
  • Ensures optimal evaporation protection during thermal cycling
    Precision Thermal Performance
  • Ultra-thin-walled wells made from 100% virgin medical-grade polypropylene
  • Uniform and thin wall thickness promotes rapid and consistent heat transfer, reducing cycle times
    Versatile Compatibility
  • Available with multiple skirt options for a secure fit in most common thermal cyclers
    Optimized for Workflow Efficiency
  • Plates are available in various colors to help distinguish workflow steps
  • Alphanumeric coding ensures easy and accurate sample identification
  • White plates are specifically designed for qPCR, providing optimal fluorescence signal reflection
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Untreated
Certifications/Compliance BIO-CERT PCR Quality
Format Standard
Lid No
Description PCR plate 384-well, rigid frame, PC/PP, frosted, wells white, Roche™ Lightcycler™, BioRad, BIO-CERT™ PCR QUALITY
Array 16 x 24
Autoclavable Autoclavable
Packaging Bagged
Color Frosted
Disposable Single-use
Product Type PCR 96-well plate
For Use With (Equipment) Roche Lightcycler, BioRad Cycler
Skirt Style Full
Material Polycarbonate, Polypropylene
Volume (Metric) Well 0.03 mL
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Dnase Free Yes
No. per Pack 50
Profile Low Profile
RNase Free Yes
Well Color White
Length (Metric) 26 cm
Width (Metric) 23 cm
Height (Metric) 9 cm
Fit for Purpose PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.