missing translation for 'onlineSavingsMsg'
Learn More

Rimantadine hydrochloride, MedChemExpress

Product Code. 16482139
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482139 1 g 1g
16448246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482139 Supplier Medchem Express Supplier No. HYB0338A
 Restricted Item
Estimated Shipment: 08-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rimantadine hydrochloride is an anti-influenza virus drug.

Rimantadine hydrochloride is an anti-influenza virus drug.

Specifications

Chemical Name or Material Rimantadine hydrochloride
Physical Form Solid
CAS 1501-84-4
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C12H22ClN
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 50 mg/mL (231.74 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (154.48 mM; Need ultrasonic)
SMILES CC(C1(C[C@H](C2)C3)C[C@H]3C[C@H]2C1)N.Cl
Molecular Weight (g/mol) 215.76
Formula Weight 215.76
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.