Learn More
RIPK1/RIP1 Antibody (CL11357), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21190-100ul
 View more versions of this product
Product Code. 18663704
  • 600.00 € / 100µL
Description
RIPK1/RIP1 Monoclonal antibody specifically detects RIPK1/RIP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RIPK1/RIP1
Monoclonal
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot
CL11357
Western Blot 1 ug/ml
Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP
This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of Q13546. Actual immunogen sequence is proprietary.
100 μg
Apoptosis, Necroptosis
8737
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a