missing translation for 'onlineSavingsMsg'
Learn More

Risedronate sodium, MedChemExpress

Product Code. 16482026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482026 500 mg 500mg
16448116 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482026 Supplier Medchem Express Supplier No. HYB0119
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Risedronate sodium is a pyridinyl biphosphonate which inhibits osteoclast-mediated bone resorption.

Risedronate sodium is a pyridinyl biphosphonate which inhibits osteoclast-mediated bone resorption.

Specifications

Chemical Name or Material Risedronate sodium
Physical Form Solid
CAS 115436-72-1
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C7H10NNaO7P2
Synonym Risedronic Acid Sodium
Solubility Information H2O : 8.33 mg/mL (27.30 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (3.28 mM; ultrasonic and warming and heat to 80°C)
SMILES OC(P(O)(O)=O)(P(O)([O-])=O)CC1=CN=CC=C1.[Na+]
Molecular Weight (g/mol) 305.09
Formula Weight 305.09
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.