missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rituximab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
337.00 € - 2465.00 €
Specifications
| Antigen | Rituximab |
|---|---|
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Hybridoma |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16408019
|
ACROBiosystems
RIB-Y37-100ug |
100 μg |
337.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16418019
|
ACROBiosystems
RIB-Y37-1mg |
1 mg |
2465.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rituximab Monoclonal antibody specifically detects Rituximab.Specifications
| Rituximab | |
| Unconjugated | |
| RUO | |
| -20°C |
| Monoclonal | |
| Hybridoma | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title