missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RMND5A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-56367-25ul
This item is not returnable.
View return policy
Description
RMND5A Polyclonal specifically detects RMND5A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| RMND5A | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| C-terminal to LisH motif, 44 kDa, CTLH, FLJ12753, FLJ13910, FLJ21795, MGC78451, p44°CTLH, protein RMD5 homolog A, required for meiotic nuclear division 5 homolog A (S. cerevisiae), RMD5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| RMND5A | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RELEKVLHKFSGYGQLCERGLEELIDYTGGLKHEILQSHGQDAELSGTLSLVLTQCCKRIKDT | |
| 25 μL | |
| Stem Cell Markers | |
| 64795 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering