missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Polymerase II/POLR2A, p Thr4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
208.00 € - 494.00 €
Specifications
| Antigen | RNA Polymerase II/POLR2A (p Thr4) |
|---|---|
| Concentration | 1.1 mg/mL |
| Dilution | Western Blot 1:1000, Dot Blot 1:1000 |
| Applications | Western Blot, Dot Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18228843
|
Novus Biologicals
NBP1-49546SS |
0.025 mL |
208.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18719203
|
Novus Biologicals
NBP1-49546 |
494.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
RNA Polymerase II/POLR2A Polyclonal specifically detects RNA Polymerase II/POLR2A in Human, Chicken, Drosophila, Yeast samples. It is validated for Western Blot, Dot Blot.Specifications
| RNA Polymerase II/POLR2A (p Thr4) | |
| Western Blot 1:1000, Dot Blot 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Chromatin Research, Epigenetics, microRNA, Phospho Specific, Transcription Factors and Regulators | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
| POLR2A | |
| IgG | |
| Affinity Purified | |
| This antibody is specific for the phosphorylated Threonine found in position 4 of the RNA polymerase II CTD heptad. |
| 1.1 mg/mL | |
| Western Blot, Dot Blot | |
| Unconjugated | |
| RUO | |
| Human, Chicken, Drosophila, Yeast | |
| P24928 | |
| 5430 | |
| A synthetic peptide made to a phosphorylated Threonine (position 4) of the CTD heptad in the human RNA polymerase II protein [UniProt P24928] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title