missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNase L Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
193.00 € - 470.00 €
Specifications
| Antigen | RNase L |
|---|---|
| Dilution | Western Blot 1:100 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693570
|
Novus Biologicals
NBP2-93434-0.02ml |
0.02 mL |
193.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677791
|
Novus Biologicals
NBP2-93434-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNase L Polyclonal antibody specifically detects RNase L in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RNase L | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase | |
| PBS (pH 7.3), 50% glycerol | |
| 6041 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 472-741 of human RNASEL (NP_066956.1). VQELHLSCGYTHQDLQPQNILIDSKKAAHLADFDKSIKWAGDPQEVKRDLEDLGRLVLYVVKKGSISFEDLKAQSNEEVVQLSPDEETKDLIHRLFHPGEHVRDCLSDLLGHPFFWTWESRYRTLRNVGNESDIKTRKSESEILRLLQPGPSEHSKSFDKWTTKINECVMKKMNKFYEKRGNFYQNTVGDLLKFIRNLGEHIDEEKHKKMKLKIGDPSLYFQKTFPDLVIYVYTKLQNTEYRKHFPQTHSPNKPQCDGAGGASGLASPGC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto