Learn More
RNASE4 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30732-1-AP-20UL
 View more versions of this product
Product Code. 18048775
  • 145.00 € / 20µL
Description
The RNASE4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RNASE4. This antibody recognizes human, mouse, rat antigen. The RNASE4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
RNASE4
350 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
Ribonuclease 4, RNase 4, RNASE4, RNS4
20 μL
6038
Store at -20°C. Stable for one year after shipment.