missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93965-0.02ml
This item is not returnable.
View return policy
Description
RNF10 Polyclonal antibody specifically detects RNF10 in Rat samples. It is validated for Western Blot
Specifications
| RNF10 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500 | |
| KIAA0262RIE2MGC126758, MGC126764, ring finger protein 10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 552-811 of human RNF10 (NP_055683.3). SMSEDVRQRHRYLSHLPLTCEFSICELALQPPVVSKETLEMFSDDIEKRKRQRQKKAREERRRERRIEIEENKKQGKYPEVHIPLENLQQFPAFNSYTCSSDSALGPTSTEGHGALSISPLSRSPGSHADFLLTPLSPTASQGSPSFCVGSLEEDSPFPSFAQMLRVGKAKADVWPKTAPKKDENSLVPPAPVDSDGESDNSDRVPVPSFQNSFSQAIEAAFMKLDTPATSDPLSEEKGGKKRKKQKQKLLFSTSVVHTK | |
| 0.02 mL | |
| Zinc Finger | |
| 9921 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction