missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF2 Recombinant Rabbit Monoclonal Antibody (24GB5455)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

198.00 € - 564.00 €

Specifications

Antigen RNF2
Clone 24GB5455
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276411
GREENER_CHOICE
View Documents
Invitrogen™
MA557696
100 μL
564.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276677
View Documents
Invitrogen™
MA557697
20 μL
198.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RNF2 Recombinant Monoclonal Antibody for Western Blot, Flow

RNF2 (E3 ubiquitin-protein ligase RING2) is a ligase that mediates monoubiquitination of Lys-119 of histone H2A (H2AK119Ub). Therefore, it plays a central role in histone code and gene regulation. The ligase activity is enhanced by BMI1/PCGF4. H2AK119Ub gives a specific tag for epigeneti transcriptional repression and participates in X chromosome inactivation of female mammals. RNF2 may be involved in the initiation of both imprinted and random X inactivation. RNF2 is also an essential component of a Polycomb group (PcG) multiprotein PRC1-like complex. This complex class is required to maintain the transcriptionally repressive state of many genes including Hox genes throughout development. The PcG PRC1 complex acts via chromatin remodeling and modification of histones and rendering chromatin heritably changed in its expressibility.
Specifications

Specifications

RNF2
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
RNF2
AI326319; AI450156; AU019207; BAP 1; BAP1; BAP-1; DinG; DinG protein; E3 ubiquitin-protein ligase RING2; fa19h05; GS1-120K12.1; HIP2-interacting protein 3; HIPI 3; HIPI3; Huntingtin-interacting protein 2-interacting protein 3; OTTHUMP00000060668; protein DinG; RING 1B; RING 2; RING finger protein 1B; ring finger protein 2; RING finger protein BAP-1; RING1B; RING2; RING-type E3 ubiquitin transferase RING2; RNF 2; Rnf2; wu:fa19h05; wz4556; wz4566
RNF2
Primary
Affinity chromatography
24GB5455
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q4KLY4, Q99496, Q9CQJ4
19821, 304850, 6045
A synthesized peptide derived from human RING2 / RING1B / RNF2 (aa 118-131).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.