missing translation for 'onlineSavingsMsg'
Learn More

Ro 41-1049 hydrochloride, MedChemExpress

Product Code. 16434216
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434216

Brand: Medchem Express HY100027A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ro 41-1049 hydrochloride is a reversible and selective inhibitor of monoamine oxidase-A (MAO-A). An homogeneous population of high affinity binding sites for [3H]Ro 41-1049 is found in membrane preparations from human frontal cortex and placenta (Kd values of 16.5 and 64.4 nM, respectively).

Ro 41-1049 hydrochloride is a reversible and selective inhibitor of monoamine oxidase-A (MAO-A). An homogeneous population of high affinity binding sites for [3H]Ro 41-1049 is found in membrane preparations from human frontal cortex and placenta (Kd values of 16.5 and 64.4 nM, respectively).

Specifications

Chemical Name or Material Ro 41-1049 hydrochloride
CAS 127917-66-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H13ClFN3OS
Quantity 100 mg
Solubility Information DMSO : ≥ 32 mg/mL (106.04 mM) ∣H2O : 25 mg/mL (82.84 mM; Need ultrasonic)
SMILES O=C(C1=C(C2=CC=CC(F)=C2)SC=N1)NCCN.[H]Cl
Molecular Weight (g/mol) 301.77
Formula Weight 301.77
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.